missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexanedione bis(ethylene acetal), 99%

Product Code. 11485676
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11485676 10 g 10g
11495676 50 g 50g
11405686 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11485676 Supplier Thermo Scientific Alfa Aesar Supplier No. B20784.09
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 183-97-1 | C10H16O4 | 200.234 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 183-97-1
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.234
MDL Number MFCD00010851
InChI Key YSMVSEYPOBXSOK-UHFFFAOYSA-N
Synonym 1,4-cyclohexanedione bis ethylene ketal, 1,4,9,12-tetraoxadispiro 4.2.4.2 tetradecane, 1,4-cyclohexanedione bis ethylene acetal, 1,4,9,12-tetraoxadispiro 4,2,4,2 tetradecane, 1,4-cyclohexanedione bis ethyleneketal, 1,4,9,12-tetraoxadispiro 4.2.4^ 8 .2^ 5 tetradecane, 1,4,9,12-tetraoxadispiro 4.2.4?.2? tetradecane, maybridge1_006651, 1,4-cyclohexanedione ethyleneketal
PubChem CID 135986
IUPAC Name 1,4,9,12-tetraoxadispiro[4.2.4^{8}.2^{5}]tetradecane
SMILES C1CC2(CCC13OCCO3)OCCO2

Specifications

Melting Point 79°C to 80°C
Boiling Point 75°C to 85°C (0.5 mmHg)
Quantity 10 g
Formula Weight 200.23
Percent Purity 99%
Chemical Name or Material 1,4-Cyclohexanedione bis(ethylene acetal)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.