missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexanedione, 98%

Product Code. 11408983
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408983 500 g 500g
11488973 25 g 25g
11498973 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11408983 Supplier Thermo Scientific Alfa Aesar Supplier No. A11100.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 637-88-7 | C6H8O2 | 112.128 g/mol

1,4-Cyclohexanedione is used in the preparation of 1,4 benzoquinone and bromoorganics. It is also used to study the influence of visible light on the bromate-1,4-cyclohexanedione-ferroin oscillating reaction. It plays a vital role in pharmaceuticals, plant growth regulator and as a conducting material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Cyclohexanedione is used in the preparation of 1,4 benzoquinone and bromoorganics. It is also used to study the influence of visible light on the bromate-1,4-cyclohexanedione-ferroin oscillating reaction. It plays a vital role in pharmaceuticals, plant growth regulator and as a conducting material.

Solubility
Soluble in methanol and ethanol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-88-7
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00001606
InChI Key DCZFGQYXRKMVFG-UHFFFAOYSA-N
Synonym 1,4-cyclohexanedione, tetrahydroquinone, 1,4-dioxocyclohexane, 1,4-cyclohexandione, 1,4 cyclohexane dione, 1,4-cyclohexane-dione, cyclohexan-1,4-dione, pubchem13686, cyclohexane-1,4-quinone, acmc-1b8ar
PubChem CID 12511
ChEBI CHEBI:28286
IUPAC Name cyclohexane-1,4-dione
SMILES C1CC(=O)CCC1=O

Specifications

Melting Point 77°C to 78°C
Boiling Point 132°C (20 mmHg)
Flash Point 132°C (269°F)
Quantity 500 g
Beilstein 774152
Solubility Information Soluble in methanol and ethanol.
Formula Weight 112.13
Percent Purity 98%
Chemical Name or Material 1,4-Cyclohexanedione

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.