Learn More
1,4-Cyclohexanediol, cis + trans, 98%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 556-48-9 |
---|---|
Molecular Formula | C6H12O2 |
Molecular Weight (g/mol) | 116.16 |
MDL Number | MFCD00001448,MFCD00063612,MFCD00075462 |
InChI Key | VKONPUDBRVKQLM-UHFFFAOYSA-N |
Synonym | 1,4-cyclohexanediol, cis-1,4-cyclohexanediol, trans-1,4-cyclohexanediol, trans-cyclohexane-1,4-diol, quinitol, 1,4-cyclohexanediol, trans, cis-cyclohexane-1,4-diol, 1,4-cyclohexanediol, cis, trans-1,4-dihydroxycyclohexane, cis-1,4-dihydroxycyclohexane |
PubChem CID | 11162 |
IUPAC Name | cyclohexane-1,4-diol |
SMILES | OC1CCC(O)CC1 |
Description
1,4-Cyclohexanediol, cis + trans, is used as a chemical, organic, pharmaceutical intermediate. It is also used as an important intermediate for manufacturing of anti-Cancer and anti-Infective Drugs - dihydroartemisinin and dihydroartemisitene dimers. It is also used in co polyester resins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,4-Cyclohexanediol, cis + trans, is used as a chemical, organic, pharmaceutical intermediate. It is also used as an important intermediate for manufacturing of anti-Cancer and anti-Infective Drugs - dihydroartemisinin and dihydroartemisitene dimers. It is also used in co polyester resins.
Solubility
It is highly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, acid chlorides, acid anhydrides, strong acids.
Chemical Identifiers
556-48-9 | |
116.16 | |
VKONPUDBRVKQLM-UHFFFAOYSA-N | |
11162 | |
OC1CCC(O)CC1 |
C6H12O2 | |
MFCD00001448,MFCD00063612,MFCD00075462 | |
1,4-cyclohexanediol, cis-1,4-cyclohexanediol, trans-1,4-cyclohexanediol, trans-cyclohexane-1,4-diol, quinitol, 1,4-cyclohexanediol, trans, cis-cyclohexane-1,4-diol, 1,4-cyclohexanediol, cis, trans-1,4-dihydroxycyclohexane, cis-1,4-dihydroxycyclohexane | |
cyclohexane-1,4-diol |
Specifications
556-48-9 | |
149°C to 151°C (20 mmHg) | |
C6H12O2 | |
2036658 | |
It is highly soluble in water. | |
OC1CCC(O)CC1 | |
116.16 | |
116.16 | |
1,4-Cyclohexanediol, cis + trans |
98°C to 103°C | |
65°C (149°F) | |
MFCD00001448,MFCD00063612,MFCD00075462 | |
1,4-cyclohexanediol, cis-1,4-cyclohexanediol, trans-1,4-cyclohexanediol, trans-cyclohexane-1,4-diol, quinitol, 1,4-cyclohexanediol, trans, cis-cyclohexane-1,4-diol, 1,4-cyclohexanediol, cis, trans-1,4-dihydroxycyclohexane, cis-1,4-dihydroxycyclohexane | |
VKONPUDBRVKQLM-UHFFFAOYSA-N | |
cyclohexane-1,4-diol | |
11162 | |
98% |
Safety and Handling
missing translation for 'einecsNumber' : 209-126-2
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only