missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Cyclohexanedimethanol, TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | 1,4-Cyclohexanedimethanol |
| CAS | 105-08-8 |
| Molecular Formula | C24 H30 O3 Si |
| Quantity | 50 g |
| Synonym | 1,4-Bis(hydroxymethyl)cyclohexane, 1,4-Cyclohexamethylenebis(methylol), 1,4-Cyclohexane dimethylol, 1,4-Dimethylolcyclohexane, CHDM, CHDM-D, Cyclohex-1,4-ylenedimethanol, NSC 44508, Rikabinol DM, Sky CHDM, Vibracure A 240 |
| InChI Formula | InChI=1S/C24H30O3Si/c1-24(2,3)28(19-10-6-4-7-11-19,20-12-8-5-9-13-20)26-17-18-14-15-22-21(18)16-23(25)27-22/h4-13,18,21-22H,14-17H2,1-3H3/t18-,21+,22+/m1/s1 |
| SMILES | CC(C)(C)[Si](OC[C@H]1CC[C@@H]2OC(=O)C[C@@H]12)(c3ccccc3)c4ccccc4 |
| IUPAC Name | (3aS,4S,6aS)-4-[[tert-butyl(diphenyl)silyl]oxymethyl]-3,3a,4,5,6,6a-hexahydrocyclopenta[b]furan-2-one |
| Molecular Weight (g/mol) | 394.579 |
| Formula Weight | 394.196 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction