missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexanedimethanol, cis + trans, 99%

Product Code. p-7023335
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11432788

Brand: Thermo Scientific Alfa Aesar A16769.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 105-08-8 | C8H16O2 | 144.21 g/mol

CHDM is a precursor to polyesters. It is one of the most important co-monomers for production of polyethylene terephthalate (PET), or polyethylene terephthalic ester (PETE).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
CHDM is a precursor to polyesters. It is one of the most important co-monomers for production of polyethylene terephthalate (PET), or polyethylene terephthalic ester (PETE).

Solubility
Miscible with water, glycolether and ketones.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-08-8
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00001512,MFCD00066360
InChI Key YIMQCDZDWXUDCA-UHFFFAOYSA-N
Synonym 1,4-cyclohexanedimethanol, trans-1,4-cyclohexanedimethanol, chdm, 1,4-bis hydroxymethyl cyclohexane, rikabinol dm, cyclohexane-1,4-dimethanol, 1,4-chidm, 1,4-dimethylolcyclohexane, 4-hydroxymethyl cyclohexyl methanol, 1,4-cyclohexanedimethanol, cis
PubChem CID 7735
IUPAC Name [4-(hydroxymethyl)cyclohexyl]methanol
SMILES OCC1CCC(CO)CC1

Specifications

Melting Point ∼30°C
Density 1.04
Boiling Point 282°C to 284°C
Flash Point 161°C (322°F)
Quantity 100 g
Beilstein 1902271
Solubility Information Miscible with water,glycolether and ketones.
Formula Weight 144.21
Percent Purity 99%
Chemical Name or Material 1,4-Cyclohexanedimethanol, cis + trans
compliance-icons
Product Identifier
  • 1,4-Cyclohexanedimethanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : 1,4-Cyclohexanedimethanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.