missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexanedimethanol, cis + trans, 99%

Product Code. 11442788
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11442788 500 g 500g
11432788 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11442788 Supplier Thermo Scientific Alfa Aesar Supplier No. A16769.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 105-08-8 | C8H16O2 | 144.21 g/mol

CHDM is a precursor to polyesters. It is one of the most important co-monomers for production of polyethylene terephthalate (PET), or polyethylene terephthalic ester (PETE).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
CHDM is a precursor to polyesters. It is one of the most important co-monomers for production of polyethylene terephthalate (PET), or polyethylene terephthalic ester (PETE).

Solubility
Miscible with water, glycolether and ketones.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-08-8
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00001512,MFCD00066360
InChI Key YIMQCDZDWXUDCA-UHFFFAOYSA-N
Synonym 1,4-cyclohexanedimethanol, trans-1,4-cyclohexanedimethanol, chdm, 1,4-bis hydroxymethyl cyclohexane, rikabinol dm, cyclohexane-1,4-dimethanol, 1,4-chidm, 1,4-dimethylolcyclohexane, 4-hydroxymethyl cyclohexyl methanol, 1,4-cyclohexanedimethanol, cis
PubChem CID 7735
IUPAC Name [4-(hydroxymethyl)cyclohexyl]methanol
SMILES OCC1CCC(CO)CC1

Specifications

Melting Point ∼30°C
Density 1.04
Boiling Point 282°C to 284°C
Flash Point 161°C (322°F)
Quantity 500 g
Beilstein 1902271
Solubility Information Miscible with water,glycolether and ketones.
Formula Weight 144.21
Percent Purity 99%
Chemical Name or Material 1,4-Cyclohexanedimethanol, cis + trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.