missing translation for 'onlineSavingsMsg'
Learn More

1,4-Cyclohexadiene, 97%, stabilized, Thermo Scientific Chemicals

Catalog Number p-3736065
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
628-41-1
C6H8
80.13
MFCD00001535
UVJHQYIOXKWHFD-UHFFFAOYSA-N
1,4-cyclohexadiene, 1,4-dihydrobenzene, 1,4-cyclohexanediene, unii-0f8z5909qz, 1,4 cyclohexadiene, acmc-209snj, ksc354m7f, 1,4-cyclohexadiene, stabilized 5g, 1,4-cyclohexadiene, purum gc
12343
CHEBI:37611
cyclohexa-1,4-diene
C1C=CCC=C1
This item is not returnable. View return policy
628-41-1
C6H8
80.13
MFCD00001535
UVJHQYIOXKWHFD-UHFFFAOYSA-N
1,4-cyclohexadiene, 1,4-dihydrobenzene, 1,4-cyclohexanediene, unii-0f8z5909qz, 1,4 cyclohexadiene, acmc-209snj, ksc354m7f, 1,4-cyclohexadiene, stabilized 5g, 1,4-cyclohexadiene, purum gc
12343
CHEBI:37611
cyclohexa-1,4-diene
C1C=CCC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point -49.2°C
Color Colorless
Density 0.8400g/mL
Boiling Point 81.0°C to 82.0°C
Flash Point −6°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4710 to 1.4730
Quantity 5 mL
Specific Gravity 0.84
Stabilizer 0.1% BHT
Formula Weight 80.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1, 4-Cyclohexadiene, Stabilized
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Cyclohexadiene, stabilized
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'