missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Cyanophenyl)-4-piperidinecarbohydrazide, 97%, Thermo Scientific™

Product Code. 10782113
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10782113 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10782113 Supplier Thermo Scientific Maybridge Supplier No. MO00066DA
 Restricted Item
Estimated Shipment: 28-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 352018-91-8
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C13H16N4O
MDL Number MFCD03086202
Quantity 1 g
Synonym 1-4-cyanophenyl piperidine-4-carbohydrazide, 1-4-cyanophenyl-4-piperidinecarbohydrazide, 4-4-???piperidyl benzenecarbonitrile
InChI Key RGOGBQRLKKDOHU-UHFFFAOYSA-N
SMILES NNC(=O)C1CCN(CC1)C1=CC=C(C=C1)C#N
IUPAC Name 1-(4-cyanophenyl)piperidine-4-carbohydrazide
Molecular Weight (g/mol) 244.30
PubChem CID 2779654
Formula Weight 244.3
Percent Purity 97%
Chemical Name or Material 1-(4-Cyanophenyl)-4-piperidinecarbohydrazide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.