missing translation for 'onlineSavingsMsg'
Learn More

1,4-Chrysenequinone, MedChemExpress

Product Code. 16450767
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16450767 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16450767 Supplier Medchem Express Supplier No. HY111441
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 1,4-Chrysenequinone, a polycyclic aromatic quinone, acts as an activator of aryl hydrocarbon receptor (AhR).

1,4-Chrysenequinone, a polycyclic aromatic quinone, acts as an activator of aryl hydrocarbon receptor (AhR).

Specifications

Chemical Name or Material 1,4-Chrysenequinone
CAS 100900-16-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H10O2
Quantity 50 mg
Synonym Chrysene-1,4-dione
Solubility Information DMSO : 3.33 mg/mL (12.89 mM; Need ultrasonic)
SMILES O=C(C1=C2C=CC3=C1C=CC4=CC=CC=C43)C=CC2=O
Molecular Weight (g/mol) 258.27
Formula Weight 258.27
Percent Purity 93.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.