missing translation for 'onlineSavingsMsg'
Learn More

1-[(4-chlorophenyl)sulfonyl]-1H-pyrrole, 97%, Thermo Scientific™

Product Code. p-2694102
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10233062 10 g 10g
10386434 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10233062 Supplier Thermo Scientific Maybridge Supplier No. BTB07116EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 16851-83-5
Molecular Formula C10H8ClNO2S
Molecular Weight (g/mol) 241.689
MDL Number MFCD00067753
InChI Key ACNNPPGRQRFTSO-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl sulfonyl-1h-pyrrole, 1-4-chlorobenzenesulfonyl pyrrole, maybridge1_001769, cbmicro_045913, 1-4-chlorophenyl sulfonylpyrrole, 1-chloro-4-pyrrolylsulfonyl benzene, 4-chloro-1-pyrrolylsulfonyl benzene, 1-4-chlorobenzenesulfonyl-1h-pyrrole, 1-4-chlorophenylsulfonyl-1h-pyrrole
PubChem CID 706366
IUPAC Name 1-(4-chlorophenyl)sulfonylpyrrole
SMILES C1=CN(C=C1)S(=O)(=O)C2=CC=C(C=C2)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 241.7
Percent Purity 97%
Chemical Name or Material 1-[(4-chlorophenyl)sulfonyl]-1H-pyrrole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.