missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)piperazine dihydrochloride, 95%, Thermo Scientific™

Product Code. p-7028139
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11450858

Brand: Thermo Scientific Alfa Aesar B25283.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38869-46-4
Molecular Formula C10H14Cl2N2
Molecular Weight (g/mol) 233.14
MDL Number MFCD00012766
InChI Key ZHGRQBSZTVJDHU-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl piperazine dihydrochloride, 1-4-chlorophenyl-piperazine dihydrochloride, 1-4-chlorophenyl piperazine 2hcl, para-chlorophenylpiperazine dihydrochloride, piperazine, 1-4-chlorophenyl-, dihydrochloride, pubchem8586, acmc-1ctdy, ksc493m4r, 1-p-chlorophenyl piperazine, 4-chlorophenylpiperazine dihydrochloride
PubChem CID 12212341
IUPAC Name 1-(4-chlorophenyl)piperazine;dihydrochloride
SMILES [H+].[Cl-].ClC1=CC=C(C=C1)N1CCNCC1

Specifications

Melting Point 275°C to 278°C (decomposition)
Quantity 5 g
Sensitivity Hygroscopic
Formula Weight 269.6
Percent Purity 95%
Chemical Name or Material 1-(4-Chlorophenyl)piperazine dihydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.