missing translation for 'onlineSavingsMsg'
Learn More

1-[(4-Chlorophenyl)phenylmethyl]piperazine, TRC

Product Code. 30135862
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135862 10 g 10g
30012762 25 g 25g
17981975 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135862 Supplier Toronto Research Chemicals Supplier No. TRCC37957510G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 303-26-4
Molecular Formula C17 H19 Cl N2
Molecular Weight (g/mol) 286.8
Synonym 1-[(RS)-(4-Chlorophenyl)phenylmethyl]piperazine, 1-(4-Chlorobenzhydryl)piperazine, Hydroxyzine Hydrochloride Imp. A (EP), Meclozine Hydrochloride Imp. H (EP), Cetirizine Dihydrochloride Imp. A (EP), 1-[(4-Chlorophenyl)phenylmethyl]piperazine
IUPAC Name 1-[(4-chlorophenyl)-phenylmethyl]piperazine
SMILES Clc1ccc(cc1)C(N2CCNCC2)c3ccccc3

Specifications

Quantity 10 g
InChI Formula InChI=1S/C17H19ClN2/c18-16-8-6-15(7-9-16)17(14-4-2-1-3-5-14)20-12-10-19-11-13-20/h1-9,17,19H,10-13H2
Formula Weight 286.1237
Chemical Name or Material 1,4-Bis(4-Chlorobenzhydryl)Piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.