missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)biguanide Hydrochloride, Mikromol™

Product Code. 17931386
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Product Code. Quantity unitSize
17931386 25 mg 25mg
1 options
This item is not returnable. View return policy

Product Code. 17931386

Brand: Mikromol MM0162.230025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1-(4-Chlorophenyl)biguanide Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(4-Chlorophenyl)biguanide Hydrochloride
CAS 4022-81-5
Analyte or Component Names 1-(4-Chlorophenyl)biguanide Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C8 H10 Cl N5 . Cl H
Certifications/Compliance ISO 9001
InChI Formula InChI=1S/C8H10ClN5.ClH/c9-5-1-3-6(4-2-5)13-8(12)14-7(10)11;/h1-4H,(H6,10,11,12,13,14);1H
SMILES Cl.NC(=N)NC(=N)Nc1ccc(Cl)cc1
IUPAC Name 1-carbamimidoyl-3-(4-chlorophenyl)guanidine;hydrochloride
Molecular Weight (g/mol) 248.11
Quantity 25 mg
Formula Weight 247.0392 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.