missing translation for 'onlineSavingsMsg'
Learn More

1-(4-chlorophenyl)-5-methyl-1H-pyrazole-4-carbonyl chloride, 97%, Thermo Scientific™

Product Code. 10091314
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10091314 5 g 5g
10242772 250 mg 250mg
10324272 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10091314 Supplier Thermo Scientific Maybridge Supplier No. CC00081DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 288252-38-0
Molecular Formula C11H8Cl2N2O
Molecular Weight (g/mol) 255.098
MDL Number MFCD03407325
InChI Key KUQHQFYKJPXTNB-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl-5-methyl-1h-pyrazole-4-carbonyl chloride, 1-4-chlorophenyl-5-methylpyrazole-4-carbonyl chloride, 1-4-chlorophenyl-5-methyl-1h-pyrazole-4-carbonylchloride, 1-4-chlorophenyl-5-methylpyrazole-4-carboxylic chloride, 1-4-chloro-phenyl-5-methyl-1h-pyrazole-4-carbonyl chloride, 1h-pyrazole-4-carbonylchloride, 1-4-chlorophenyl-5-methyl
PubChem CID 2776128
IUPAC Name 1-(4-chlorophenyl)-5-methylpyrazole-4-carbonyl chloride
SMILES CC1=C(C=NN1C2=CC=C(C=C2)Cl)C(=O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 255.1
Percent Purity 97%
Chemical Name or Material 1-(4-chlorophenyl)-5-methyl-1H-pyrazole-4-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.