missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)-1H-1,2,3-triazole-4-methanol, 98%, Thermo Scientific™

Product Code. 15489116
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133902-66-6
Molecular Formula C9H8ClN3O
Molecular Weight (g/mol) 209.633
MDL Number MFCD00140928
InChI Key CAHIFLPAMJOAGI-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl-1h-1,2,3-triazol-4-yl methanol, 1-4-chlorophenyl triazol-4-yl methanol, 1-4-chlorophenyl-1,2,3-triazol-4-yl methanol, 1-4-chlorophenyl-1h-1,2,3-triazole-4-yl-methanol, 1-4-chlorophenyl-1,2,3-triazol-4-yl methan-1-ol, pubchem13003, chlorophenyltriazolylmethanol, 1-4-chlorophenyl-1h-1,2,3-triazol-4-ylmethanol
PubChem CID 2763878
IUPAC Name [1-(4-chlorophenyl)triazol-4-yl]methanol
SMILES C1=CC(=CC=C1N2C=C(N=N2)CO)Cl

Specifications

Melting Point 141°C to 145°C
Quantity 250 mg
Beilstein 4250389
Formula Weight 209.63
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 1-(4-Chlorophenyl)-1H-1,2,3-triazole-4-methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'