missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)-1-cyclopentanecarboxylic acid, 98%, Thermo Scientific™

Product Code. 10327390 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10327390 25 g Glass bottle 25g
10235830 5 g Glass bottle 5g
10578863 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10327390 Supplier Thermo Scientific Acros Supplier No. 161570250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 80789-69-1
Molecular Formula C12H13ClO2
Molecular Weight (g/mol) 224.69
MDL Number MFCD00001373
InChI Key QJNFJEMGWIQMJT-UHFFFAOYSA-N
Synonym 1-4-chlorophenyl cyclopentanecarboxylic acid, 1-4-chlorophenyl-1-cyclopentanecarboxylic acid, 1-4-chlorophenyl cyclopentane-1-carboxylic acid, cyclopentanecarboxylic acid, 1-4-chlorophenyl, asischem d50961, timtec-bb sbb000649, acmc-209pk2, labotest-bb lt00453384, 1-4-chloro phenyl cyclopentanecarboxylic acid, 1-4-chloro-phenyl-cyclopentylcarboxylic acid
PubChem CID 97447
IUPAC Name 1-(4-chlorophenyl)cyclopentane-1-carboxylic acid
SMILES C1CCC(C1)(C2=CC=C(C=C2)Cl)C(=O)O

Specifications

Melting Point 160.0°C to 164.0°C
CAS Min % 97.5
Color Cream to Gray
CAS Max % 100.0
Quantity 25 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder or Crystals
Assay Percent Range 98%
Linear Formula ClC6H4C5H8CO2H
Formula Weight 224.69
Percent Purity 98%
Chemical Name or Material 1-(4-Chlorophenyl)-1-cyclopentanecarboxylic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.