1-(4-Chlorophenyl)-1-cyclopentanecarboxylic acid, 98%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 161570050

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 80789-69-1 Weight: 0.05500kg


Packaging Glass bottle
Quantity 5g

Product Code. 10235830

Quantity Price
1 £ 27.7 / 5g
Estimated Shipment date
from Supplier 13-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 1-(4-Chlorophenyl)-1-cyclopentanecarboxylic acid
CAS 80789-69-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C12H13ClO2
Linear Formula ClC6H4C5H8CO2H
Formula Weight 224.69
MDL Number MFCD00001373

Specification

Physical Form Crystalline Powder or Crystals
Assay ≥97.5%
Formula Weight 224.69
Quantity 5g
Color Cream to gray
CAS Min % 97.5
Linear Formula ClC6H4C5H8CO2H
CAS 80789-69-1
Infrared Spectrum Authentic
Assay Percent Range 98%
MDL Number MFCD00001373
Packaging Glass bottle
Molecular Formula C12H13ClO2
CAS Max % 100.0
Chemical Name or Material 1-(4-Chlorophenyl)-1-cyclopentanecarboxylic acid
Melting Point 160° to 165°C
Documentation

Technical documents