missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)-1-cyclohexanecarboxylic acid, 95%

Product Code. 10723561 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10723561 5 g Glass bottle 5g
10730681 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10723561 Supplier Thermo Scientific Acros Supplier No. 170760050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 58880-37-8 | C13H14ClO2 | 237.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58880-37-8
Molecular Formula C13H14ClO2
Molecular Weight (g/mol) 237.70
MDL Number MFCD00019350
InChI Key UPNXUJXIIZGXLQ-UHFFFAOYSA-M
Synonym 1-4-chlorophenyl cyclohexanecarboxylic acid, 1-4-chlorophenyl cyclohexane-1-carboxylic acid, 1-4-chlorophenyl-1-cyclohexanecarboxylic acid, 1-4-chlorophenyl-1-cyclohexane-carboxylic acid, cyclohexanecarboxylic acid, 1-4-chlorophenyl, acmc-1cuiq, 4-chlorophenylcyclohexanecarboxylic acid, 4-chlorophenyl cyclohexanecarboxylic acid, 1-4-chlorophenyl cyclohexanecarboxylicacid
PubChem CID 100873
SMILES [O-]C(=O)C1(CCCCC1)C1=CC=C(Cl)C=C1

Specifications

Melting Point 150°C to 156°C
CAS Min % 94.0
Color Gray
CAS Max % 100.0
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Powder and Granules
Assay Percent Range 95%
Linear Formula ClC6H4(C6H10)COOH
IUPAC Name 1-(4-chlorophenyl)cyclohexane-1-carboxylic acid
Formula Weight 238.71
Percent Purity 95%
Chemical Name or Material 1-(4-Chlorophenyl)-1-cyclohexanecarboxylic acid, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.