missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorophenyl)-1-cyclohexanecarbonitrile, 99%, Thermo Scientific™

Product Code. 10532581 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
This item is not returnable. View return policy

Product Code. 10532581

Brand: Thermo Scientific Acros 170650025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Melting Point 88.0°C to 92.0°C
CAS 64399-28-6
CAS Min % 98.5
Color White to Beige
CAS Max % 100.0
Quantity 2.5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Assay Percent Range 98.5% min. (GC)
Molecular Formula C13H14ClN
MDL Number MFCD00065289
Synonym 1-4-chlorophenyl cyclohexanecarbonitrile, 1-4-chlorophenyl-1-cyclohexanecarbonitrile, 1-4-chlorophenyl cyclohexane-1-carbonitrile, cyclohexanecarbonitrile, 1-4-chlorophenyl, 1-4-chlorophenyl-cyclohexanecarbonitrile, 1-4-chlorophenyl cyclohexanecarbonitrile #, cyclohexanecarbonitrile,1-4-chlorophenyl, 1-4-chlorophenyl-1-cyclohexane-carbonitrile
InChI Key OSHOYPXOCQTDGJ-UHFFFAOYSA-N
SMILES C1CCC(CC1)(C#N)C2=CC=C(C=C2)Cl
IUPAC Name 1-(4-chlorophenyl)cyclohexane-1-carbonitrile
Molecular Weight (g/mol) 219.71
PubChem CID 116613
Formula Weight 219.71
Percent Purity 99%
Chemical Name or Material 1-(4-Chlorophenyl)-1-cyclohexanecarbonitrile, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 1-(4-Chlorophenyl)-1-cyclohexanecarbonitrile
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.