missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Chlorophenyl)-1,5-dihydro-4-(4-morpholinyl)-2H-imidazol-2-one, TRC
missing translation for 'orderingAttributeHoverText'
Quantity:
50 mg
missing translation for 'unitSize'
50mg
Specifications
Specifications
| Chemical Name or Material | 1-(4-Chlorophenyl)-1,5-dihydro-4-(4-morpholinyl)-2H-imidazol-2-one |
| CAS | 188116-07-6 |
| Molecular Formula | C13 H14 Cl N3 O2 |
| Quantity | 50 mg |
| Synonym | 1-(4-Chlorophenyl)-1,5-dihydro-4-(4-morpholinyl)-2H-imidazol-2-one |
| InChI Formula | InChI=1S/C13H14ClN3O2/c14-10-1-3-11(4-2-10)17-9-12(15-13(17)18)16-5-7-19-8-6-16/h1-4H,5-9H2 |
| SMILES | Clc1ccc(cc1)N2CC(=NC2=O)N3CCOCC3 |
| IUPAC Name | 3-(4-chlorophenyl)-5-morpholin-4-yl-4H-imidazol-2-one |
| Molecular Weight (g/mol) | 279.722 |
| Formula Weight | 279.077 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction