missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorobenzyl)piperazine, 98%

Product Code. 15461558
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15461558 5 g 5g
15471558 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15461558 Supplier Thermo Scientific Alfa Aesar Supplier No. H55337.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 23145-88-2 | C11H15ClN2 | 210.705 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23145-88-2
Molecular Formula C11H15ClN2
Molecular Weight (g/mol) 210.705
MDL Number MFCD00040791
InChI Key GSJXJZOWHSTWOX-UHFFFAOYSA-N
Synonym 1-4-chlorobenzyl piperazine, 1-4-chlorophenyl methyl piperazine, 4-chlorobenzylpiperazine, 1-4-chlorobenzyl-piperazine, 1-4-chloro-benzyl-piperazine, 1-4-chloro benzyl piperazine, n-4-chlorobenzyl piperazine, 4-chlorophenyl methyl piperazine, norhomochlorcyclizine, chlorobenzylpiperazin
PubChem CID 134826
IUPAC Name 1-[(4-chlorophenyl)methyl]piperazine
SMILES C1CN(CCN1)CC2=CC=C(C=C2)Cl

Specifications

Color Colorless to Yellow
Density 1.140 g/mL
Boiling Point 104°C to 106°C (0.1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.558
Quantity 5 g
UN Number UN3267
Formula Weight 210.71
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-(4-Chlorobenzyl)piperazine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.