missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorobenzyl)piperazine 98%, ACROS Organics™

Product Code. p-3593204 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10288373

Brand: Thermo Scientific Acros 391950010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 23145-88-2
Molecular Formula C11H15ClN2
Molecular Weight (g/mol) 210.705
MDL Number MFCD00040791
InChI Key GSJXJZOWHSTWOX-UHFFFAOYSA-N
Synonym 1-4-chlorobenzyl piperazine, 1-4-chlorophenyl methyl piperazine, 4-chlorobenzylpiperazine, 1-4-chlorobenzyl-piperazine, 1-4-chloro-benzyl-piperazine, 1-4-chloro benzyl piperazine, n-4-chlorobenzyl piperazine, 4-chlorophenyl methyl piperazine, norhomochlorcyclizine, chlorobenzylpiperazin
PubChem CID 134826
IUPAC Name 1-[(4-chlorophenyl)methyl]piperazine
SMILES C1CN(CCN1)CC2=CC=C(C=C2)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.1400g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5560 to 1.5580 (20°C, 589nm)
Quantity 1 g
Formula Weight 210.71
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 1-(4-Chlorobenzyl)piperazine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.