missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorobenzyl)piperazine 98%, ACROS Organics™

Product Code. 10444515 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10444515 5 g Glass bottle 5g
10288373 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10444515 Supplier Thermo Scientific Acros Supplier No. 391950050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 23145-88-2
Molecular Formula C11H15ClN2
Molecular Weight (g/mol) 210.705
MDL Number MFCD00040791
InChI Key GSJXJZOWHSTWOX-UHFFFAOYSA-N
Synonym 1-4-chlorobenzyl piperazine, 1-4-chlorophenyl methyl piperazine, 4-chlorobenzylpiperazine, 1-4-chlorobenzyl-piperazine, 1-4-chloro-benzyl-piperazine, 1-4-chloro benzyl piperazine, n-4-chlorobenzyl piperazine, 4-chlorophenyl methyl piperazine, norhomochlorcyclizine, chlorobenzylpiperazin
PubChem CID 134826
IUPAC Name 1-[(4-chlorophenyl)methyl]piperazine
SMILES C1CN(CCN1)CC2=CC=C(C=C2)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.1400g/mL
Boiling Point 104.0°C to 106.0°C (0.1 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5560 to 1.5580
Quantity 5 g
Formula Weight 210.71
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-(4-Chlorobenzyl)piperazine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-(4-Chlorobenzyl)piperazine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.