missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Chlorobenzhydryl)piperazine, 90-95%

Product Code. 10656482 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10656482 5 g Glass bottle 5g
10489780 25 g Glass bottle 25g
11946511 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10656482 Supplier Thermo Scientific Acros Supplier No. 108650050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 303-26-4 | C17H19ClN2 | 286.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 303-26-4
Molecular Formula C17H19ClN2
Molecular Weight (g/mol) 286.79
InChI Key UZKBSZSTDQSMDR-UHFFFAOYSA-N
Synonym 1-4-chlorobenzhydryl piperazine, norchlorcyclizine, 1-4-chlorophenyl phenyl methyl piperazine, n-p-chlorobenzhydryl piperazine, 4-4-chlorobenzhydryl piperazine, 1-4-chlorophenyl phenylmethyl piperazine, n-4-chlorobenzhydryl piperazine, 1-4-chlorophenyl-phenylmethyl piperazine
PubChem CID 9340
IUPAC Name 1-[(4-chlorophenyl)-phenylmethyl]piperazine
SMILES C1CN(CCN1)C(C2=CC=CC=C2)C3=CC=C(C=C3)Cl

Specifications

Melting Point 65.0°C to 75.0°C
Color White to Beige
Boiling Point 178.0°C to 180.0°C (0.5 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 90% min. (on dry substance) (Tertiary base)
Packaging Glass bottle
Quantity 5 g
Formula Weight 286.79
Percent Purity 90 to 95%
Physical Form Crystalline Powder
Chemical Name or Material 1-(4-Chlorobenzhydryl)piperazine
Show More Show Less
compliance-icons
Product Identifier
  • 1-(4-Chlorobenzhydryl)piperazine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.