missing translation for 'onlineSavingsMsg'
Learn More

1,4 Butanediol, MP Biomedicals™

Product Code. 11415865 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
4 L
4 x 4 L
6 x 1 L
Unit Size:
16 L
4 L
6 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11415865 4 x 4 L 16 L
11435416 4 L 4 L
11471135 6 x 1 L 6 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11415865 Supplier MP Biomedicals Supplier No. 0230002316
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

1,4 Butanediol Certified Grade, Clear Oily Liquid

  • 1,4 Butanediol and its derivatives are used in the manufacturing of technical plastics, polyurethanes, solvents, electronic chemicals and elastic fibers.

Chemical Identifiers

CAS 110-63-4
Molecular Weight (g/mol) 90.12
MDL Number MFCD00002968
Synonym Butane-1,4-diol, 1,4-Butyleneglycol, 1,4-Dihdyroxybutane, Tetramethylene 1,4-diol

Specifications

Melting Point 16°C (literature)
Color ≤10 (APHA)
Density 1.017g/mL at 25°C (Lit.)
Boiling Point 230°C (Literature)
Flash Point 110°C / 273.2°F (Lit.)
Molecular Formula HO(CH2)4OH
Refractive Index n20/D 1.445 (Lit.)
Quantity 4 x 4 L
Beilstein 1633445
Vapor Density 3.1g/L (vs air)(Lit.)
Vapor Pressure < 1 hPa at 20°C (Lit.)
Grade Certified
Physical Form Oily Liquid
Assay ≥99.0%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.