missing translation for 'onlineSavingsMsg'
Learn More

1,4-Butanediol diacrylate, 85+%, stab. with 50-105 ppm hydroquinone

Product Code. 11323018
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11323018 25 g 25g
11313018 5 g 5g
11303018 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11323018 Supplier Thermo Scientific Alfa Aesar Supplier No. 032780.14
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1070-70-8 | C10H14O4 | 198.22 g/mol

1,4-Butanediol diacrylate is used as a multifunctional crosslinker. It is used in drying technology for paintings, coatings, inks, resins and adhesives. It serves as an intermediate product for polymer synthesis in the chemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1070-70-8
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.22
MDL Number MFCD00014940
InChI Key JHWGFJBTMHEZME-UHFFFAOYSA-N
Synonym 1,4-butanediol diacrylate, butylene diacrylate, butanediol diacrylate, tetramethylene glycol diacrylate, 1,4-butylene glycol diacrylate, tetramethylene acrylate, tetramethylene diacrylate, 1,4-butylene diacrylate, 1,4-butanediyl diacrylate, 1,4-butanediyl bisacrylate
PubChem CID 70613
SMILES C=CC(=O)OCCCCOC(=O)C=C

Specifications

Color Colorless
Density 1.051 g/mL
Boiling Point 83°C (0.3 mmHg)
Flash Point >110°C (230°F)
Odor Pleasant
Linear Formula CH2=CHCO2(CH2)4O2CCH=CH2
Refractive Index 1.456
Quantity 25 g
UN Number UN1760
Beilstein 1777487
Solubility Information Immiscible with water.
IUPAC Name 4-prop-2-enoyloxybutyl prop-2-enoate
Formula Weight 198.22
Percent Purity ≥85%
Physical Form Liquid
Assay (Reactive Esters)
Chemical Name or Material 1,4-Butanediol diacrylate, Stabilized with 50 to 105 ppm hydroquinone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.