missing translation for 'onlineSavingsMsg'
Learn More

1,4-Butane diisothiocyanate, 98%

Product Code. 11317306
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11317306 5 g 5g
11307306 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317306 Supplier Thermo Scientific Alfa Aesar Supplier No. L09234.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4430-51-7 | C6H8N2S2 | 172.264 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4430-51-7
Molecular Formula C6H8N2S2
Molecular Weight (g/mol) 172.264
MDL Number MFCD00041136
InChI Key RRSISCMPUAGVJN-UHFFFAOYSA-N
Synonym 1,4-butane diisothiocyanate, butane, 1,4-diisothiocyanato, tetramethylene diisothiocyanate, #
PubChem CID 138238
IUPAC Name 1,4-diisothiocyanatobutane
SMILES C(CCN=C=S)CN=C=S

Specifications

Melting Point 30°C to 33°C
Boiling Point 170°C (11 mmHg)
Odor Irritating
Quantity 5 g
UN Number UN2923
Beilstein 1757138
Sensitivity Moisture sensitive
Formula Weight 172.27
Percent Purity 98%
Chemical Name or Material 1,4-Butane diisothiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.