missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Bromophenyl)pyrrolidine, 97%, Thermo Scientific™

Product Code. 10239942
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
This item is not returnable. View return policy

Product Code. 10239942

Brand: Thermo Scientific Maybridge CC30110CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 22090-26-2
Molecular Formula C10H12BrN
Molecular Weight (g/mol) 226.117
InChI Key BVEGBJXZICCEQW-UHFFFAOYSA-N
Synonym 1-4-bromophenyl pyrrolidine, n-4-bromophenyl pyrrolidine, 1-4-bromophenyl-pyrrolidine, acmc-209frt, 4-pyrrolidinobromobenzene, 4-bromophenyl pyrrolidine, 1-4-bromo-phenyl-pyrrolidine, pyrrolidine,1-4-bromophenyl, 4-bromo-1-1-pyrrolidinyl benzene
PubChem CID 7016457
IUPAC Name 1-(4-bromophenyl)pyrrolidine
SMILES C1CCN(C1)C2=CC=C(C=C2)Br

Specifications

Assay Percent Range 97%
Quantity 250 mg
Formula Weight 226.12
Percent Purity 97%
Chemical Name or Material 1-(4-Bromophenyl)pyrrolidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.