Learn More
1-(4-Bromophenyl)-3,5-dimethyl-1H-pyrazole, 97%, Maybridge™
Brand: Maybridge CC68210DA
Code : 31
Additional Details : CAS Number : 62546-27-4 Weight : 0.05100kg
Packaging | Amber Glass Bottle |
---|---|
Quantity | 1g |
Chemical Identifiers
62546-27-4 | |
251.127 | |
ATCMNDCOEJOOSF-UHFFFAOYSA-N | |
24229761 | |
CC1=CC(=NN1C2=CC=C(C=C2)Br)C |
C11H11BrN2 | |
MFCD09817558 | |
1-4-bromophenyl-3,5-dimethyl-1h-pyrazole, 1-4-bromophenyl-3,5-dimethylpyrazole, 1h-pyrazole, 1-4-bromophenyl-3,5-dimethyl, acmc-209n4v | |
1-(4-bromophenyl)-3,5-dimethylpyrazole |
Specifications
1-(4-Bromophenyl)-3, 5-dimethyl-1H-pyrazole | |
1g | |
96.5 | |
97% | |
MFCD09817558 | |
ATCMNDCOEJOOSF-UHFFFAOYSA-N | |
1-(4-bromophenyl)-3,5-dimethylpyrazole | |
24229761 | |
97% |
Amber Glass Bottle | |
62546-27-4 | |
100.0 | |
C11H11BrN2 | |
1-4-bromophenyl-3,5-dimethyl-1h-pyrazole, 1-4-bromophenyl-3,5-dimethylpyrazole, 1h-pyrazole, 1-4-bromophenyl-3,5-dimethyl, acmc-209n4v | |
CC1=CC(=NN1C2=CC=C(C=C2)Br)C | |
251.127 | |
251.13 |