missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Bromophenoxy)hexane, 97%, Thermo Scientific™

Product Code. 10382283
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10382283 10 g 10g
10679214 1 g 1g
10128852 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10382283 Supplier Thermo Scientific Maybridge Supplier No. BTB09795EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 30752-19-3
Molecular Formula C12H17BrO
Molecular Weight (g/mol) 257.171
InChI Key GKLMJONYGGTHHM-UHFFFAOYSA-N
Synonym 1-bromo-4-hexyloxy benzene, 1-bromo-4-hexyloxybenzene, 1-4-bromophenoxy hexane, 1-bromo-4-n-hexyloxybenzene, 4-n-hexyloxybromobenzene, p-bromophenylhexyl ether, benzene, 1-bromo-4-hexyloxy, p-hexyloxybromobenzene, 4-hexyloxybromobenzene, 4-bromophenyl hexyl ether
PubChem CID 520492
IUPAC Name 1-bromo-4-hexoxybenzene
SMILES CCCCCCOC1=CC=C(C=C1)Br

Specifications

Assay Percent Range 97%
Quantity 10 g
Formula Weight 257.17
Chemical Name or Material 1-(4-Bromophenoxy)hexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.