missing translation for 'onlineSavingsMsg'
Learn More

1-[4-(Bromomethyl)phenyl]-1H-1,2,4-triazole, 97%, Thermo Scientific™

Product Code. 10304142
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10304142 1 g Amber Glass Bottle 1g
10009231 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10304142 Supplier Thermo Scientific Maybridge Supplier No. CC18608DA
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 58419-69-5
Molecular Formula C9H8BrN3
Molecular Weight (g/mol) 238.088
MDL Number MFCD04113600
InChI Key NOUAEJFAVOHNSY-UHFFFAOYSA-N
Synonym 1-4-bromomethyl phenyl-1h-1,2,4-triazole, 1-4-bromomethyl phenyl-1,2,4-triazole, 1h-1,2,4-triazole, 1-4-bromomethyl phenyl, 1-4-bromomethylphenyl-1h-1,2,4-triazole, zlchem 1040, 1-4-bromomethyl phenyl-1h-1,2,4-triazol, 1-4-bromomethyl-phenyl-1h-1,2,4-triazole, 1-4-bromo-methyl phenyl-1h-1,2,4-triazole
PubChem CID 2776489
IUPAC Name 1-[4-(bromomethyl)phenyl]-1,2,4-triazole
SMILES C1=CC(=CC=C1CBr)N2C=NC=N2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 319
Percent Purity 97%
Chemical Name or Material 1-[4-(Bromomethyl)phenyl]-1H-1, 2, 4-triazole hydrobromide hydrate
compliance-icons
Product Identifier
  • 1-[4-(Bromomethyl)phenyl]-1H-1,2,4-triazole hydrobromide
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.