missing translation for 'onlineSavingsMsg'
Learn More

1-[4-(Bromomethyl)phenyl]-1H-1,2,4-triazole, 97%, Maybridge™

Brand:  Maybridge CC18608CB

Code : 31

Additional Details : CAS Number : 58419-69-5 Weight : 0.05025kg Transport : UN number : 1759 Chem class : 8 Pack group : III


Packaging Amber Glass Bottle
Quantity 250mg

 View more versions of this product

Product Code. 10009231

  • £77.35 / 250mg

Estimated Shipment
01-06-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

58419-69-5
238.088
NOUAEJFAVOHNSY-UHFFFAOYSA-N
2776489
C1=CC(=CC=C1CBr)N2C=NC=N2
C9H8BrN3
MFCD04113600
1-4-bromomethyl phenyl-1h-1,2,4-triazole, 1-4-bromomethyl phenyl-1,2,4-triazole, 1h-1,2,4-triazole, 1-4-bromomethyl phenyl, 1-4-bromomethylphenyl-1h-1,2,4-triazole, zlchem 1040, 1-4-bromomethyl phenyl-1h-1,2,4-triazol, 1-4-bromomethyl-phenyl-1h-1,2,4-triazole, 1-4-bromo-methyl phenyl-1h-1,2,4-triazole
1-[4-(bromomethyl)phenyl]-1,2,4-triazole

Specifications

1-[4-(Bromomethyl)phenyl]-1H-1, 2, 4-triazole hydrobromide hydrate
250mg
96.5
97%
MFCD04113600
NOUAEJFAVOHNSY-UHFFFAOYSA-N
1-[4-(bromomethyl)phenyl]-1,2,4-triazole
2776489
97%
Amber Glass Bottle
58419-69-5
100.0
C9H8BrN3
1-4-bromomethyl phenyl-1h-1,2,4-triazole, 1-4-bromomethyl phenyl-1,2,4-triazole, 1h-1,2,4-triazole, 1-4-bromomethyl phenyl, 1-4-bromomethylphenyl-1h-1,2,4-triazole, zlchem 1040, 1-4-bromomethyl phenyl-1h-1,2,4-triazol, 1-4-bromomethyl-phenyl-1h-1,2,4-triazole, 1-4-bromo-methyl phenyl-1h-1,2,4-triazole
C1=CC(=CC=C1CBr)N2C=NC=N2
238.088
319
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 1-¢4-(Bromomethyl)phenyl!-1H-1,2,4-triazole hydrobromide

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS Documents