missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Bromobenzyl)-5-methyl-1H-1,2,3-triazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10022464
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10022464 5 g 5g
10278812 250 mg 250mg
10288812 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10022464 Supplier Thermo Scientific Maybridge Supplier No. MO07389DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 845885-94-1
Molecular Formula C11H10BrN3O2
Molecular Weight (g/mol) 296.124
MDL Number MFCD06659088
InChI Key FJFFHBYCFJNIAV-UHFFFAOYSA-N
Synonym 1-4-bromobenzyl-5-methyl-1h-1,2,3-triazole-4-carboxylic acid, 1-4-bromophenyl methyl-5-methyl-1,2,3-triazole-4-carboxylic acid, 1-4-bromophenyl methyl-5-methyl-1h-1,2,3-triazole-4-carboxylic acid, 1h-1,2,3-triazole-4-carboxylicacid, 1-4-bromophenyl methyl-5-methyl, 1-4-bromophenyl methyl-5-methyl-4-triazolecarboxylic acid, 1-4-bromophenyl methyl-5-methyltriazole-4-carboxylic acid, 1-4-bromo-benzyl-5-methyl-1h-1,2,3 triazole-4-carboxylic acid
PubChem CID 2794828
IUPAC Name 1-[(4-bromophenyl)methyl]-5-methyltriazole-4-carboxylic acid
SMILES CC1=C(N=NN1CC2=CC=C(C=C2)Br)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 296.12
Percent Purity 97%
Chemical Name or Material 1-(4-Bromobenzyl)-5-methyl-1H-1, 2, 3-triazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.