missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Bromobenzyl)-4-methylperhydro-1,4-diazepine, 97%, Thermo Scientific™

Product Code. 10422664
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10422664 1 g Amber Glass Bottle 1g
11356795 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10422664 Supplier Thermo Scientific Maybridge Supplier No. CC55810DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 280560-78-3
Molecular Formula C13H19BrN2
Molecular Weight (g/mol) 283.213
MDL Number MFCD00810550
InChI Key WQUWRVLHCKJAMG-UHFFFAOYSA-N
Synonym 1-4-bromobenzyl-4-methylperhydro-1,4-diazepine, 1-4-bromophenyl methyl-4-methyl-1,4-diazepane, 1-4-bromobenzyl-4-methyl-1,4-diazepane, cambridge id 5258520, 1h-1,4-diazepine,1-4-bromophenyl methyl hexahydro-4-methyl
PubChem CID 763047
IUPAC Name 1-[(4-bromophenyl)methyl]-4-methyl-1,4-diazepane
SMILES CN1CCCN(CC1)CC2=CC=C(C=C2)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 283.21
Percent Purity 97%
Chemical Name or Material 1-(4-Bromobenzyl)-4-methylperhydro-1, 4-diazepine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.