missing translation for 'onlineSavingsMsg'
Learn More

1-(4-bromobenzyl)-1,4-diazepane, 95%, Thermo Scientific™

Catalog Number 10539722
Click to view available options
Amber Glass Bottle; 1g
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 690632-73-6
CAS Min % 97.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C12H17BrN2
MDL Number MFCD03407498
Quantity 1 g
Synonym 1-4-bromobenzyl-1,4-diazepane, 1-4-bromophenyl methyl-1,4-diazepane, 1-b-bromophenyl methyl-1,4-diazepane, 1h-1,4-diazepine,1-4-bromophenyl methyl hexahydro
InChI Key PUTBARDRYLXING-UHFFFAOYSA-N
SMILES BrC1=CC=C(CN2CCCNCC2)C=C1
IUPAC Name 1-[(4-bromophenyl)methyl]-1,4-diazepane
Molecular Weight (g/mol) 269.19
PubChem CID 2794807
Formula Weight 269.18
Percent Purity 95%
Chemical Name or Material 1-(4-Bromobenzyl)-1, 4-diazepane
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'