missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Bromo-2-fluorobenzyl)piperazine, 96%, Thermo Scientific™

Product Code. 15482208
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 870703-75-6
Molecular Formula C11H14BrFN2
Molecular Weight (g/mol) 273.15
MDL Number MFCD07368744
InChI Key NRCBEINBLSTDHM-UHFFFAOYSA-N
Synonym 1-4-bromo-2-fluorobenzyl piperazine, 1-4-bromo-2-fluorophenyl methyl piperazine, acmc-20apb9
PubChem CID 2782804
IUPAC Name 1-[(4-bromo-2-fluorophenyl)methyl]piperazine
SMILES FC1=C(CN2CCNCC2)C=CC(Br)=C1

Specifications

Color Yellow to Orange
Density 1.411 g/mL
Boiling Point 120°C to 122°C (0.02 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.563
Quantity 1 g
Formula Weight 273.15
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 1-(4-Bromo-2-fluorobenzyl)piperazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'