missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Bromo-2-fluorobenzyl)piperazine, 96%

Brand:  Alfa Aesar™ H55844.14

Code : NEW

Additional Details : CAS Number : 870703-75-6 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15402218

  • £461.00 / 25g

Estimated Shipment: 26-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

870703-75-6
273.15
NRCBEINBLSTDHM-UHFFFAOYSA-N
2782804
FC1=C(CN2CCNCC2)C=CC(Br)=C1
C11H14BrFN2
MFCD07368744
1-4-bromo-2-fluorobenzyl piperazine, 1-4-bromo-2-fluorophenyl methyl piperazine, acmc-20apb9
1-[(4-bromo-2-fluorophenyl)methyl]piperazine

Specifications

1-(4-Bromo-2-fluorobenzyl)piperazine
>110°C (230°F)
120°C to 122°C (0.02mmHg)
25g
C11H14BrFN2
1-4-bromo-2-fluorobenzyl piperazine, 1-4-bromo-2-fluorophenyl methyl piperazine, acmc-20apb9
FC1=C(CN2CCNCC2)C=CC(Br)=C1
273.15
273.15
96%
1.411 g/mL
1.563
Yellow to Orange
870703-75-6
MFCD07368744
NRCBEINBLSTDHM-UHFFFAOYSA-N
1-[(4-bromo-2-fluorophenyl)methyl]piperazine
2782804
Liquid
Videos
SDS Documents
Certificates