missing translation for 'onlineSavingsMsg'
Learn More

1-(4-bromo-1,3-thiazol-2-yl)-4-methylpiperazine, 97%, Thermo Scientific™

Product Code. 10642675
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10642675 5 g Amber Glass Bottle 5g
10402074 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10642675 Supplier Thermo Scientific Maybridge Supplier No. CC77310DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 919352-66-2
Molecular Formula C8H12BrN3S
Molecular Weight (g/mol) 262.17
MDL Number MFCD09878991
InChI Key GHLARJPRKBAWMZ-UHFFFAOYSA-N
Synonym 1-4-bromo-1,3-thiazol-2-yl-4-methylpiperazine, 4-bromo-2-4-methylpiperazin-1-yl thiazole, 4-bromo-2-4-methylpiperazin-1-yl-1,3-thiazole, 1-4-bromo-thiazol-2-yl-4-methyl-piperazine, 4-bromo-2-4-methylpiperazinyl-1,3-thiazole, piperazine, 1-4-bromo-2-thiazolyl-4-methyl, 2-4-methylpiperazin-1-yl-4-bromothiazole, 4-bromo-2-n-methylpiperazin-1-yl thiazole
PubChem CID 43811061
IUPAC Name 4-bromo-2-(4-methylpiperazin-1-yl)-1,3-thiazole
SMILES CN1CCN(CC1)C1=NC(Br)=CS1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 262.17
Percent Purity 97%
Chemical Name or Material 1-(4-Bromo-1, 3-thiazol-2-yl)-4-methylpiperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.