missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Boc-1-piperidinyl)-4-bromopyrazole, 98%, Thermo Scientific Chemicals

Product Code. 15442519
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 877399-50-3
Molecular Formula C13H20BrN3O2
Molecular Weight (g/mol) 330.23
MDL Number MFCD09831980
InChI Key IYNZAVDBHAQODX-UHFFFAOYSA-N
Synonym tert-butyl 4-4-bromo-1h-pyrazol-1-yl piperidine-1-carboxylate, 1-boc-4-4-bromopyrazol-1-yl piperidine, 4-4-bromopyrazol-1-yl piperidine-1-carboxylic acid tert-butyl ester, 4-4-bromo-pyrazol-1-yl-piperidine-1-carboxylic acid tert-butyl ester, tert-butyl 4-4-bromopyrazol-1-yl piperidine-1-carboxylate, n-boc-4-4-bromopyrazol-1-yl piperidine, 3-2-ethoxycarbonylacetyl piperidine-1-carboxylic acid tert-butyl ester, 1-piperidinecarboxylicacid, 4-4-bromo-1h-pyrazol-1-yl-, 1,1-dimethylethyl ester, ksc658s5d, 1-boc-4-4-bromo-1-pyrazolyl piperidine
PubChem CID 45480278
IUPAC Name tert-butyl 4-(4-bromopyrazol-1-yl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)N1C=C(Br)C=N1

Specifications

Color White
Quantity 250 mg
Beilstein 14511017
Formula Weight 330.23
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 1-(4-Boc-1-piperidinyl)-4-bromopyrazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'