missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis[(trimethylsilyl)ethynyl]benzene, 98%

Product Code. 11322227
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11322227 5 g 5g
11312227 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322227 Supplier Thermo Scientific Alfa Aesar Supplier No. L09709.06
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 17938-13-5 | C16H22Si2 | 270.522 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17938-13-5
Molecular Formula C16H22Si2
Molecular Weight (g/mol) 270.522
MDL Number MFCD00135433
InChI Key CMTMWEXUJQSPCA-UHFFFAOYSA-N
Synonym 1,4-bis trimethylsilyl ethynyl benzene, 1,4-bis trimethylsilylethynyl benzene, 1,4-bis 2-trimethylsilyl ethynyl benzene, trimethyl-2-4-2-trimethylsilylethynyl phenyl ethynyl silane, trimethyl 2-4-2-trimethylsilyl ethynyl phenyl ethynyl silane, acmc-20ap2w, trimethyl 4-trimethylsilyl ethynyl phenyl ethynyl silane, benzene,1,4-bis 2-trimethylsilyl ethynyl
PubChem CID 522492
IUPAC Name trimethyl-[2-[4-(2-trimethylsilylethynyl)phenyl]ethynyl]silane
SMILES C[Si](C)(C)C#CC1=CC=C(C=C1)C#C[Si](C)(C)C

Specifications

Melting Point 119°C to 121°C
Quantity 5 g
Beilstein 2840618
Formula Weight 270.53
Percent Purity 98%
Chemical Name or Material 1,4-Bis[(trimethylsilyl)ethynyl]benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.