missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(trimethylsilyl)-1,3-butadiyne, 98%

Product Code. 11387306
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387306 1 g 1g
11397306 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387306 Supplier Thermo Scientific Alfa Aesar Supplier No. L09246.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4526-07-2 | C10H18Si2 | 194.42 g/mol

1,4-Bis(trimethylsilyl)-1,3-butadiyne is used in Negishi protocol for the synthesis of glycosylated oligo(ethynylene)s. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Bis(trimethylsilyl)-1,3-butadiyne is used in Negishi protocol for the synthesis of glycosylated oligo(ethynylene)s. It is also used as pharmaceutical intermediates.

Solubility
Insoluble in water.

Notes
Store at 4°C. Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4526-07-2
Molecular Formula C10H18Si2
Molecular Weight (g/mol) 194.42
MDL Number MFCD00009839
InChI Key LBNVCJHJRYJVPK-UHFFFAOYSA-N
SMILES C[Si](C)(C)C#CC#C[Si](C)(C)C

Specifications

Melting Point 110°C to 113°C
Quantity 1 g
UN Number UN1325
Beilstein 1758268
Synonym 3-methyl-4-isopropylaniline hydrochloride, 4-isopropyl-3-methylaniline hydrochloride, 4-isopropyl-m-toluidinium chloride, benzenamine, 3-methyl-4-1-methylethyl-, hydrochloride, acmc-20a42e, 3-methyl-4-propan-2-ylaniline hydrochloride, 4-isopropyl-3-methylbenzenamine hydrochloride, 3-methyl-4-methylethyl phenylamine, chloride, 3-methyl-4-propan-2-yl aniline hydrochloride, benzenamine, 3-?methyl-?4-? 1-?methylethyl ?-?, hy
IUPAC Name 3-methyl-4-propan-2-ylaniline;hydrochloride
PubChem CID 2796270
Formula Weight 194.43
Percent Purity 98%
Chemical Name or Material 1,4-Bis(trimethylsilyl)-1,3-butadiyne

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.