missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(trifluoromethyl)benzene, 99%

Product Code. 11494220
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494220 100 g 100g
11478337 5 g 5g
11488337 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494220 Supplier Thermo Scientific Alfa Aesar Supplier No. A18411.22
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 433-19-2 | C8H4F6 | 214.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 433-19-2
Molecular Formula C8H4F6
Molecular Weight (g/mol) 214.11
MDL Number MFCD00000402
InChI Key PDCBZHHORLHNCZ-UHFFFAOYSA-N
Synonym 1,4-bis trifluoromethyl benzene, 1,4-di trifluoromethyl benzene, 1,4-bis trifluoromethyl-benzene, hexafluoro-p-xylene, p-bis trifluoromethyl benzene, benzene, 1,4-bis trifluoromethyl, 1,4-bis-trifluoromethylbenzene, p-trifluoromethylbenzotrifluoride, ptf-tfm, alpha,alpha,alpha,alpha',alpha',alpha'-hexafluoro-p-xylene
PubChem CID 67945
IUPAC Name 1,4-bis(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point -1°C
Density 1.381
Boiling Point 116°C to 117°C
Flash Point 21°C (69°F)
Odor Aromatic
Refractive Index 1.379
Quantity 100 g
UN Number UN1993
Beilstein 1912445
Formula Weight 214.11
Percent Purity 99%
Chemical Name or Material 1,4-Bis(trifluoromethyl)benzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.