missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(trichloromethyl)benzene, 99%, Thermo Scientific™

Product Code. p-68864 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10615592

Brand: Thermo Scientific Acros 154170010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-36-0
Molecular Formula C8H4Cl6
Molecular Weight (g/mol) 312.82
MDL Number MFCD00000791
InChI Key OTEKOJQFKOIXMU-UHFFFAOYSA-N
Synonym 1,4-bis trichloromethyl benzene, bitriben, chloksil, chloxil, chloxyl, hexichol, khloksil, khloxil, cloxil, hetol
PubChem CID 6233
IUPAC Name 1,4-bis(trichloromethyl)benzene
SMILES ClC(Cl)(Cl)C1=CC=C(C=C1)C(Cl)(Cl)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 106°C to 110°C
Color Yellow
Boiling Point 312°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H4(CCl3)2
Quantity 1 g
Beilstein 05, 385
Merck Index 15, 1304
Solubility Information Solubility in water: insoluble
Viscosity 2.07 mPa.s (120°C)
Formula Weight 312.83
Percent Purity 99%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 1, 4-Bis(trichloromethyl)benzene, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 1,4-Bis(trichloromethyl)benzene
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.