missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(p-tolylamino)anthracene-9,10-dione, TRC

Product Code. 30414719
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30414719 250 mg -
30413998 500 mg -
30410627 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30414719 Supplier Toronto Research Chemicals Supplier No. TRCP768855250MG
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128-80-3
Molecular Formula C28 H22 N2 O2
Molecular Weight (g/mol) 418.49
Synonym 1,4-Bis[(4-methylphenyl)amino]-9,10-anthracenedione, 1,4-Bis(4'-methylanilino)anthraquinone, 1,4-Bis(p-tolylamino)anthraquinone, 1,4-Bis-p-toluidino-9,10-anthraquinone, 1,4-Bis[(4-methylphenyl)amino]anthraquinone, 1,4-Di-(4'-toluidino)anthraquinone, 1,4-Di-p-toluidinoanthraquinone, 11091 Green, Ahcoquinone Cyanine Green Base, Alizarine Cyanine Green Base, Alizarine Cyanine Green G Fat Soluble, Alizarine Green G Base, Amaplast Green OZ, Anthraquinone Green G Base, Arlosol Green B, Arlosol Green BS, Arlosol Green BSS, Brown 921, C-Green 10, C.I. 61565, C.I. Disperse Green 6:1, C.I. Solvent Green 3, Ceres Green BB, Cyanine Green G base, D 4, D 4 (dye), D and C Green No. 6, D&C Green 6, DC Green 6, Diaresin Green C, FD and C Green no. 6, Fat Soluble Anthraquinone Green, Fat Soluble Green Anthraquinone, Green 6, Green No. 202, Japan Green 202, KP Plast Green G, Kayaset Green A-B, Macrolex Green 5B, NSC 84207, Nitro Fast Green GB, Nitro Fast Green GSB, Oil Green 530, Oil Green 5602, Oplas Green 533, Organol Fast Green J, Organol Green J, Orient Oil Green 530, Quinazarin green, Quinizarin Green SS, Quinizarine Green Base, Sandoplast Green GSB, Solvent Green 3, Sudan Green 4B, Sumiplast Green G, Toyo Oriental Oil Blue G, Transparent Green 5B, Waxoline Green 6GFW, Waxoline Green G
IUPAC Name 1,4-bis(4-methylanilino)anthracene-9,10-dione
SMILES Cc1ccc(Nc2ccc(Nc3ccc(C)cc3)c4C(=O)c5ccccc5C(=O)c24)cc1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C28H22N2O2/c1-17-7-11-19(12-8-17)29-23-15-16-24(30-20-13-9-18(2)10-14-20)26-25(23)27(31)21-5-3-4-6-22(21)28(26)32/h3-16,29-30H,1-2H3
Formula Weight 418.1681
Chemical Name or Material Solvent Green 3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.