missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(chloromethyl)-2,5-dimethoxybenzene, 97%, Thermo Scientific™

Product Code. 10466862
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10466862

Brand: Thermo Scientific Maybridge RJC03902DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 3752-97-4
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C10H12Cl2O2
Linear Formula (CH2Cl)2C6H2(OCH3)2
MDL Number MFCD00018881
Quantity 1 g
Synonym 1,4-bis chloromethyl-2,5-dimethoxybenzene, 2,5-bis chloromethyl-1,4-dimethoxybenzene, benzene, 1,4-bis chloromethyl-2,5-dimethoxy, 1,4-bis chlormethyl-2,5-dimethoxybenzene, 1,4-bis-chlormethyl-2,5-dimethoxybenzene
InChI Key GEUWDCJVDLHSNQ-UHFFFAOYSA-N
SMILES COC1=CC(=C(C=C1CCl)OC)CCl
IUPAC Name 1,4-bis(chloromethyl)-2,5-dimethoxybenzene
Molecular Weight (g/mol) 235.104
PubChem CID 77357
Formula Weight 235.11
Percent Purity 97%
Chemical Name or Material 1, 4-Bis(chloromethyl)-2, 5-dimethoxybenzene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.