missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis[alpha-(4-chlorophenyl)benzyl]piperazine dihydrochloride, 97%

Product Code. 15448606
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15448606

Brand: Thermo Scientific Alfa Aesar H63961.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 346451-15-8 | C30H30Cl4N2 | 560.384 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 346451-15-8
Molecular Formula C30H30Cl4N2
Molecular Weight (g/mol) 560.384
MDL Number MFCD03844641
InChI Key DLEQNCKNBPYHEY-UHFFFAOYSA-N
Synonym 1,4-bis 4-chlorophenyl phenylmethyl piperazine dihydrochloride, cetirizine impurity d, 1,4-bis 4-chlorophenyl phenylmethyl-piperazine dihydrochloride, 1,4-bis 4-chlorophenyl phenyl methyl piperazine dihydrochloride, 1,4-bis 4-chlorophenyl-phenylmethyl piperazine;dihydrochloride
PubChem CID 24720891
IUPAC Name 1,4-bis[(4-chlorophenyl)-phenylmethyl]piperazine;dihydrochloride
SMILES C1CN(CCN1C(C2=CC=CC=C2)C3=CC=C(C=C3)Cl)C(C4=CC=CC=C4)C5=CC=C(C=C5)Cl.Cl.Cl

Specifications

Color White
Quantity 1 g
Formula Weight 560.38
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1,4-Bis[alpha-(4-chlorophenyl)benzyl]piperazine dihydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.