missing translation for 'onlineSavingsMsg'
Learn More

6-Chloropyridine-2-carboxaldehyde, 97%

Product Code. 15491189
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15491189

Brand: Thermo Scientific Alfa Aesar H64129.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54087-03-5 | C6H4ClNO | 141.554 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54087-03-5
Molecular Formula C6H4ClNO
Molecular Weight (g/mol) 141.554
MDL Number MFCD09832941
InChI Key XTRLIKXVRGWTKW-UHFFFAOYSA-N
Synonym 6-chloropicolinaldehyde, 6-chloro-2-pyridinecarbaldehyde, 6-chloro-pyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde, 6-chloro, 2-chloro-6-formylpyridine, 6-chloro-2-formylpyridine, 2-chloropyridine-6-carboxaldehyde, 6-chloro-2-pyridinecarboxaldehyde, pubchem13323, chloropyridinecarbaldehyde
PubChem CID 10796848
IUPAC Name 6-chloropyridine-2-carbaldehyde
SMILES C1=CC(=NC(=C1)Cl)C=O

Specifications

Melting Point 69°C to 70°C
Color White
Quantity 5 g
Beilstein 1446583
Formula Weight 141.55
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 6-Chloropyridine-2-carboxaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.