missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(2-hydroxyethyl)piperazine, 98%

Product Code. 15481188
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15481188 5 g 5g
15491188 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15481188 Supplier Thermo Scientific Alfa Aesar Supplier No. H55063.06
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 122-96-3 | C8H18N2O2 | 174.244 g/mol

It is applied for desulfurization, decarburization extraction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied for desulfurization, decarburization extraction.

Solubility
Soluble in water.

Notes
Store at 4°C. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-96-3
Molecular Formula C8H18N2O2
Molecular Weight (g/mol) 174.244
MDL Number MFCD00006157
InChI Key VARKIGWTYBUWNT-UHFFFAOYSA-N
Synonym 1,4-piperazinediethanol, 2,2'-piperazine-1,4-diyl diethanol, 1,4-bis 2-hydroxyethyl piperazine, n,n'-bis 2-hydroxyethyl piperazine, 1,4-di 2-hydroxyethyl piperazine, n,n'-dihydroxyethylpiperazine, n,n'-di 2-hydroxyethyl piperazine, unii-eq349pkx8y, piperazine, n,n'-bis 2-hydroxyethyl, 1,4-bis beta-hydroxyethyl piperazine
PubChem CID 67151
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanol
SMILES C1CN(CCN1CCO)CCO

Specifications

Melting Point 133°C to 136°C
Boiling Point 215°C to 220°C (50 mmHg)
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 174.24
Percent Purity 99%
Chemical Name or Material 1,4-Bis(2-hydroxyethyl)piperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.