missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis(1-hydroxycyclohexyl)-1,3-butadiyne, 98+%

Product Code. 11323186
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11323186 25 g 25g
11313186 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11323186

Brand: Thermo Scientific Alfa Aesar L12492.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5768-10-5 | C9H8O4 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5768-10-5
MDL Number MFCD00021401

Specifications

Melting Point 174°C to 175°C
Molecular Formula C16H22O2
Quantity 25 g
Beilstein 1246867
Synonym bis 1-hydroxycyclohexyl butadiyne, 1,4-bis 1-hydroxycyclohexyl-1,3-butadiyne, cyclohexanol, 1,1'-butadiynylenedi, 1,1'-butadiynylenedicyclohexanol, cyclohexanol, 1,1'-1,3-butadiyne-1,4-diyl bis, 1-4-1-hydroxycyclohexyl buta-1,3-diyn-1-yl cyclohexan-1-ol, acmc-1aky0, cambridge id 5106496, cyclohexanol,1'-butadiynylenedi, 6-aminobenzoic acid propyl ester
InChI Key KJOFBMLTQUHDCL-UHFFFAOYSA-N
SMILES C1CCC(CC1)(C#CC#CC2(CCCCC2)O)O
IUPAC Name 1-[4-(1-hydroxycyclohexyl)buta-1,3-diynyl]cyclohexan-1-ol
Molecular Weight (g/mol) 246.35
PubChem CID 79835
Formula Weight 246.35
Percent Purity ≥98%
Chemical Name or Material 1,4-Bis(1-hydroxycyclohexyl)-1,3-butadiyne
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.