missing translation for 'onlineSavingsMsg'
Learn More

1,4-benzodioxane-6-boronic acid, 97%, Thermo Scientific™

Product Code. 10766915
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 517874-21-4
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C14H19BO4
MDL Number MFCD05663890
Quantity 1 g
Synonym 2-2,3-dihydrobenzo b 1,4 dioxin-6-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,4-benzodioxane-6-boronic acid, pinacol ester, 2-2,3-dihydro-1,4-benzodioxin-6-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,4-benzodioxane-6-boronic acid pinacol ester, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1,4-benzodioxane, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydrobenzo-1,4-dioxoline, amtb908, 2,3-dihydro-1,4-benzodioxine-6-boronic acid, pinacol ester, 2,3-dihydrobenzo b 1,4 dioxin-6-yl boronic acid pinacol ester, 6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2,3-dihydro-benzo 1,4 dioxine
InChI Key WIIBAHCYWOUFRM-UHFFFAOYSA-N
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)OCCO3
IUPAC Name 2-(2,3-dihydro-1,4-benzodioxin-6-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol) 262.112
PubChem CID 2760592
Formula Weight 262.11
Percent Purity 97%
Chemical Name or Material 1, 4-Benzodioxane-6-boronic acid, pinacol ester
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'